Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
HTATIP2
Entrez Gene ID
539276
Gene Name
HIV-1 Tat interactive protein 2
Gene Aliases
-
Chromosome Location
Chr.29: 24565832 - 24580239 on Build ARS-UCD1.2
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001040563.1 NP_001035653.1 4 - 5 555 144
GenBank mRNA BC104577.1 - 4 - 5 555 144

View Full Product Details