Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
RFC5
Entrez Gene ID
508389
Gene Name
replication factor C subunit 5
Gene Aliases
-
Chromosome Location
Chr.17: 57183195 - 57194572 on Build ARS-UCD1.2
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001075358.1 NP_001068826.1 4 - 5 439 74
GenBank mRNA BC108105.1 - 4 - 5 439 74

View Full Product Details