Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
SDCBP
Entrez Gene ID
510979
Gene Name
syndecan binding protein
Gene Aliases
-
Chromosome Location
Chr.14: 24710921 - 24764191 on Build ARS-UCD1.2
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001075483.2 NP_001068951.1 6 - 7 752 132
GenBank mRNA BC103011.1 - 6 - 7 753 132
BT030535.1 - 7 - 8 798 132

View Full Product Details