Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
ROPN1
Entrez Gene ID
527583
Gene Name
rhophilin associated tail protein 1
Gene Aliases
-
Chromosome Location
Chr.1: 68349821 - 68384919 on Build ARS-UCD1.2
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001075900.1 NP_001069368.1 3 - 4 494 76
GenBank mRNA BC102549.1 - 3 - 4 494 76

View Full Product Details