Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
GSG1
Entrez Gene ID
530502
Gene Name
germ cell associated 1
Gene Aliases
-
Chromosome Location
Chr.5: 96766276 - 96785809 on Build ARS-UCD1.2
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001075933.1 NP_001069401.1 5 - 6 931 67
GenBank mRNA BC102722.1 - 5 - 6 931 67

View Full Product Details