Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
PLEKHO2
Entrez Gene ID
613696
Gene Name
pleckstrin homology domain containing O2
Gene Aliases
PLEKHQ1
Chromosome Location
Chr.10: 45233899 - 45256216 on Build ARS-UCD1.2
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001076283.2 NP_001069751.1 4 - 5 490 66
GenBank mRNA BC109475.1 - 4 - 5 491 66
BT026221.1 - 2 - 3 181 66

View Full Product Details