Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
MORN2
Entrez Gene ID
616607
Gene Name
MORN repeat containing 2
Gene Aliases
-
Chromosome Location
Chr.11: 21205701 - 21213907 on Build ARS-UCD1.2
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001076428.1 NP_001069896.1 4 - 5 414 86
GenBank mRNA BC109927.1 - 4 - 5 414 86

View Full Product Details