Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
C22H3orf22
Entrez Gene ID
618236
Gene Name
chromosome 22 C3orf22 homolog
Gene Aliases
C3H3orf22, C3orf22
Chromosome Location
Chr.22: 60446134 - 60451884 on Build ARS-UCD1.2
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001076497.2 NP_001069965.1 3 - 4 345 60
GenBank mRNA BC108183.1 - 3 - 4 346 60

View Full Product Details