Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
COMMD3
Entrez Gene ID
767926
Gene Name
COMM domain containing 3
Gene Aliases
-
Chromosome Location
Chr.13: 23425884 - 23429678 on Build ARS-UCD1.2
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001076999.1 NP_001070467.1 1 - 2 157 136
GenBank mRNA BC103075.1 - 1 - 2 157 136

View Full Product Details