Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
HOATZ
Entrez Gene ID
767960
Gene Name
HOATZ cilia and flagella associated protein
Gene Aliases
C15H11orf88
Chromosome Location
Chr.15: 21813732 - 21853037 on Build ARS-UCD1.2
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001077028.2 NP_001070496.1 1 - 2 230 87
GenBank mRNA BC108238.1 - 1 - 2 232 87

View Full Product Details