Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
KLHL36
Entrez Gene ID
768061
Gene Name
kelch like family member 36
Gene Aliases
C16orf44, C18H16orf44
Chromosome Location
Chr.18: 10837381 - 10853535 on Build ARS-UCD1.2
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001077118.1 NP_001070586.1 2 - 3 219 94
GenBank mRNA BC107546.1 - 2 - 3 219 94

View Full Product Details