Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
RAP1B
Entrez Gene ID
327708
Gene Name
RAP1B, member of RAS oncogene family
Gene Aliases
-
Chromosome Location
Chr.5: 45156910 - 45200871 on Build ARS-UCD1.2
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_175824.2 NP_787018.1 6 - 7 663 142
GenBank mRNA BC102330.1 - 6 - 7 663 142
M33141.1 - 5 - 6 467 142

View Full Product Details