Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
SPAM1
Entrez Gene ID
353352
Gene Name
sperm adhesion molecule 1 (PH-20 hyaluronidase, zona pellucida binding)
Gene Aliases
PH-20
Chromosome Location
Chr.4: 88200511 - 88222320 on Build ARS-UCD1.2
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001008413.3 NP_001008413.3 2 - 3 1255 107
GenBank mRNA AY297029.1 - 1 - 2 959 107
AY781776.1 - 2 - 3 1179 107
BC110183.1 - 2 - 3 1253 107

View Full Product Details