Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
CCDC65
Entrez Gene ID
535207
Gene Name
coiled-coil domain containing 65
Gene Aliases
-
Chromosome Location
Chr.5: 30885448 - 30893650 on Build ARS-UCD1.2
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001038166.2 NP_001033255.1 1 - 2 309 102
GenBank mRNA BC111168.1 - 1 - 2 310 102

View Full Product Details