Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
ILF2
Entrez Gene ID
539760
Gene Name
interleukin enhancer binding factor 2
Gene Aliases
-
Chromosome Location
Chr.3: 16711945 - 16722477 on Build ARS-UCD1.2
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001038187.2 NP_001033276.1 9 - 10 729 77
GenBank mRNA BC111202.1 - 9 - 10 730 77

View Full Product Details