Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
SPATA6L
Entrez Gene ID
613587
Gene Name
spermatogenesis associated 6 like
Gene Aliases
C8H9orf68
Chromosome Location
Chr.8: 39858145 - 39959303 on Build ARS-UCD1.2
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
GenBank mRNA BC111296.1 - 2 - 3 290 102

View Full Product Details