Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
COMMD9
Entrez Gene ID
510280
Gene Name
COMM domain containing 9
Gene Aliases
-
Chromosome Location
Chr.15: 66742642 - 66755667 on Build ARS-UCD1.2
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001038520.2 NP_001033609.1 2 - 3 200 134
GenBank mRNA BC109885.1 - 2 - 3 202 134

View Full Product Details