Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
HAUS1
Entrez Gene ID
528378
Gene Name
HAUS augmin like complex subunit 1
Gene Aliases
CCDC5
Chromosome Location
Chr.24: 45853686 - 45866281 on Build ARS-UCD1.2
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001038557.2 NP_001033646.1 5 - 6 635 96
GenBank mRNA BC110011.1 - 5 - 6 636 96

View Full Product Details