Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
ZPBP
Entrez Gene ID
654401
Gene Name
zona pellucida binding protein
Gene Aliases
-
Chromosome Location
Chr.4: 5809705 - 5961463 on Build ARS-UCD1.2
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001038611.2 NP_001033700.1 3 - 4 540 66
GenBank mRNA BC109835.1 - 3 - 4 541 66
DQ317530.1 - 2 - 3 183 66

View Full Product Details