Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
NUDT22
Entrez Gene ID
533578
Gene Name
nudix hydrolase 22
Gene Aliases
-
Chromosome Location
Chr.29: 42531948 - 42534793 on Build ARS-UCD1.2
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001038680.1 NP_001033769.1 4 - 5 723 60
GenBank mRNA BC110129.1 - 4 - 5 723 60

View Full Product Details