Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
C5H12orf45
Entrez Gene ID
616791
Gene Name
chromosome 5 C12orf45 homolog
Gene Aliases
C12orf45
Chromosome Location
Chr.5: 68528021 - 68537404 on Build ARS-UCD1.2
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001038689.2 NP_001033778.1 1 - 2 149 87
GenBank mRNA BC110124.1 - 1 - 2 150 87

View Full Product Details