Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
RCAN3
Entrez Gene ID
506601
Gene Name
RCAN family member 3
Gene Aliases
DSCR1L2
Chromosome Location
Chr.2: 128384053 - 128426795 on Build ARS-UCD1.2
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001045945.1 NP_001039410.1 1 - 2 83 107
GenBank mRNA BC112714.1 - 1 - 2 83 107

View Full Product Details