Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
MKRN2
Entrez Gene ID
508004
Gene Name
makorin ring finger protein 2
Gene Aliases
-
Chromosome Location
Chr.22: 56543251 - 56581386 on Build ARS-UCD1.2
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001045989.2 NP_001039454.1 1 - 2 87 57
GenBank mRNA BC110201.1 - 1 - 2 88 57
BT025382.1 - 1 - 2 87 57

View Full Product Details