Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
PDCD2
Entrez Gene ID
512079
Gene Name
programmed cell death 2
Gene Aliases
-
Chromosome Location
Chr.9: 104110948 - 104116975 on Build ARS-UCD1.2
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001046109.2 NP_001039574.1 1 - 2 354 105
GenBank mRNA BC110283.1 - 1 - 2 355 105

View Full Product Details