Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
GTF2B
Entrez Gene ID
513231
Gene Name
general transcription factor IIB
Gene Aliases
-
Chromosome Location
Chr.3: 55014549 - 55045664 on Build ARS-UCD1.2
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001046142.2 NP_001039607.1 2 - 3 195 76
GenBank mRNA BC112570.1 - 2 - 3 196 76

View Full Product Details