Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
RCN3
Entrez Gene ID
522073
Gene Name
reticulocalbin 3
Gene Aliases
-
Chromosome Location
Chr.18: 55968593 - 55990135 on Build ARS-UCD1.2
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001046260.1 NP_001039725.1 4 - 5 678 61
GenBank mRNA BC105535.1 - 4 - 5 678 61

View Full Product Details