Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
RELL2
Entrez Gene ID
529327
Gene Name
RELT like 2
Gene Aliases
-
Chromosome Location
Chr.7: 52730247 - 52734389 on Build ARS-UCD1.2
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001046303.1 NP_001039768.1 6 - 7 1267 79
GenBank mRNA BC112736.1 - 6 - 7 1267 79

View Full Product Details