Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
GIMAP7
Entrez Gene ID
530031
Gene Name
GTPase, IMAP family member 7
Gene Aliases
-
Chromosome Location
Chr.4: 112956771 - 112995894 on Build ARS-UCD1.2
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001046308.1 NP_001039773.1 1 - 2 207 113
GenBank mRNA BC112493.1 - 1 - 2 207 113

View Full Product Details