Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
SLC22A5
Entrez Gene ID
541266
Gene Name
solute carrier family 22 member 5
Gene Aliases
-
Chromosome Location
Chr.7: 22033881 - 22060149 on Build ARS-UCD1.2
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001046502.2 NP_001039967.1 6 - 7 1232 96
GenBank mRNA BC105377.1 - 6 - 7 1234 96

View Full Product Details