Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
HAVCR1
Entrez Gene ID
613999
Gene Name
hepatitis A virus cellular receptor 1
Gene Aliases
-
Chromosome Location
Chr.7: 68202227 - 68212987 on Build ARS-UCD1.2
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001046523.2 NP_001039988.1 3 - 4 624 145
GenBank mRNA BC112627.1 - 3 - 4 625 145

View Full Product Details