Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
TMCO5B
Entrez Gene ID
513290
Gene Name
transmembrane and coiled-coil domains 5B
Gene Aliases
-
Chromosome Location
Chr.10: 29343216 - 29359943 on Build ARS-UCD1.2
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001075609.2 NP_001069077.1 10 - 11 803 171
GenBank mRNA BC109751.1 - 10 - 11 804 171

View Full Product Details