Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
CCDC172
Entrez Gene ID
522573
Gene Name
coiled-coil domain containing 172
Gene Aliases
C26H10orf96
Chromosome Location
Chr.26: 36749674 - 36807371 on Build ARS-UCD1.2
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001075833.2 NP_001069301.1 2 - 3 182 80
GenBank mRNA BC110215.1 - 2 - 3 183 80

View Full Product Details