Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
POU2AF1
Entrez Gene ID
528475
Gene Name
POU class 2 homeobox associating factor 1
Gene Aliases
BOB-1
Chromosome Location
Chr.15: 21635821 - 21660814 on Build ARS-UCD1.2
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001075915.1 NP_001069383.1 3 - 4 330 57
GenBank mRNA BC105441.1 - 3 - 4 330 57

View Full Product Details