Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
SPAG4
Entrez Gene ID
618468
Gene Name
sperm associated antigen 4
Gene Aliases
-
Chromosome Location
Chr.13: 64840371 - 64845761 on Build ARS-UCD1.2
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001076507.2 NP_001069975.1 3 - 4 610 71
GenBank mRNA BC109514.1 - 3 - 4 611 71

View Full Product Details