Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
FAM228A
Entrez Gene ID
768001
Gene Name
family with sequence similarity 228 member A
Gene Aliases
C11H2orf84
Chromosome Location
Chr.11: 74934683 - 74946757 on Build ARS-UCD1.2
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001077061.2 NP_001070529.1 5 - 6 788 126
GenBank mRNA BC109980.1 - 5 - 6 790 126

View Full Product Details