Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
PIGF
Entrez Gene ID
768015
Gene Name
phosphatidylinositol glycan anchor biosynthesis class F
Gene Aliases
-
Chromosome Location
Chr.11: 29028850 - 29060490 on Build ARS-UCD1.2
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001077074.2 NP_001070542.1 4 - 5 484 113
GenBank mRNA BC111281.1 - 4 - 5 485 113

View Full Product Details