Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
TEX35
Entrez Gene ID
768066
Gene Name
testis expressed 35
Gene Aliases
C16H1orf49, C1orf49
Chromosome Location
Chr.16: 59914953 - 59972612 on Build ARS-UCD1.2
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001077123.2 NP_001070591.1 6 - 7 420 89
GenBank mRNA BC110123.1 - 6 - 7 421 89

View Full Product Details