Assay Design

The assay is designed to a sequence that does not exhibit typical exon-intron-exon gene structure, such as a fusion transcript, or an artificial, prokaryotic or mitochondrial sequence.

Gene Transcripts

Gene Symbol
INIP
Entrez Gene ID
616784
Gene Name
INTS3 and NABP interacting protein
Gene Aliases
C8H9orf80, C9orf80, SSBIP1
Chromosome Location
Chr.8: 101961347 - 101990100 on Build ARS-UCD1.2
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001078064.2 NP_001071532.1 - 94 80
GenBank mRNA BC111656.1 - - 95 80

View Full Product Details