The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_001079510.2 | NP_001072978.1 | 3 - 4 | 509 | 85 |
GenBank mRNA | BC110210.1 | - | 3 - 4 | 510 | 85 |