Assay Design

The amplified product spans an exon junction and the probe and one of the primers sit within one exon. This assay may detect gDNA.

Gene Transcripts

Gene Symbol
ITGB3BP
Entrez Gene ID
614469
Gene Name
integrin subunit beta 3 binding protein
Gene Aliases
-
Chromosome Location
Chr.3: 82052235 - 82157908 on Build ARS-UCD1.2
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001079619.2 NP_001073087.1 8 - 8 618 146
GenBank mRNA BC112592.1 - 8 - 8 619 146

View Full Product Details