Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
ODF3
Entrez Gene ID
509757
Gene Name
outer dense fiber of sperm tails 3
Gene Aliases
-
Chromosome Location
Chr.11: 82862083 - 82866378 on Build ARS-UCD1.2
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001080252.2 NP_001073721.1 4 - 5 1104 96
GenBank mRNA BC110233.1 - 4 - 5 1105 96

View Full Product Details