Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
TIMP3
Entrez Gene ID
282094
Gene Name
TIMP metallopeptidase inhibitor 3
Gene Aliases
TIMP-3
Chromosome Location
Chr.5: 71402333 - 71459972 on Build ARS-UCD1.2
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_174473.4 NP_776898.2 3 - 4 602 76
GenBank mRNA AB091678.1 - 2 - 3 136 76
AF144765.1 - 3 - 4 293 76
BC112663.1 - 3 - 4 603 76
BC142426.1 - 3 - 4 561 76
U77588.1 - 3 - 4 652 76

View Full Product Details