Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
YBX1
Entrez Gene ID
287023
Gene Name
Y-box binding protein 1
Gene Aliases
NSEP1, YB1
Chromosome Location
Chr.3: 103520065 - 103539625 on Build ARS-UCD1.2
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_174815.2 NP_777240.1 1 - 2 307 131
GenBank mRNA BC105363.1 - 1 - 2 307 131
M95793.1 - 1 - 2 279 131

View Full Product Details