Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
YBX1
Entrez Gene ID
287023
Gene Name
Y-box binding protein 1
Gene Aliases
NSEP1, YB1
Chromosome Location
Chr.3: 103520065 - 103539625 on Build ARS-UCD1.2
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_174815.2 NP_777240.1 5 - 6 799 73
GenBank mRNA BC105363.1 - 5 - 6 799 73
M95793.1 - 5 - 6 771 73

View Full Product Details