Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
NDUFS8
Entrez Gene ID
287027
Gene Name
NADH:ubiquinone oxidoreductase core subunit S8
Gene Aliases
-
Chromosome Location
Chr.29: 45559234 - 45563284 on Build ARS-UCD1.2
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001302669.1 NP_001289598.1 2 - 3 130 120
GenBank mRNA BC109906.1 - 2 - 3 262 120
DT719975.1 - 2 - 3 104 120
DV906691.1 - 2 - 3 131 120
M58717.1 - 2 - 3 108 120

View Full Product Details