Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
RSAD2
Entrez Gene ID
506415
Gene Name
radical S-adenosyl methionine domain containing 2
Gene Aliases
-
Chromosome Location
Chr.11: 90038987 - 90056291 on Build ARS-UCD1.2
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001045941.1 NP_001039406.1 1 - 2 398 62
GenBank mRNA BC105455.1 - 1 - 2 398 62

View Full Product Details