Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
CGREF1
Entrez Gene ID
507586
Gene Name
cell growth regulator with EF-hand domain 1
Gene Aliases
-
Chromosome Location
Chr.11: 72519739 - 72536221 on Build ARS-UCD1.2
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001045977.2 NP_001039442.1 4 - 5 249 58
GenBank mRNA BC114163.1 - 4 - 5 250 58

View Full Product Details