Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
PDCD5
Entrez Gene ID
509123
Gene Name
programmed cell death 5
Gene Aliases
TFAR19
Chromosome Location
Chr.18: 43083826 - 43090563 on Build ARS-UCD1.2
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001046024.1 NP_001039489.1 3 - 4 207 112
GenBank mRNA BC105370.1 - 3 - 4 207 112

View Full Product Details