Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
CTDNEP1
Entrez Gene ID
509192
Gene Name
CTD nuclear envelope phosphatase 1
Gene Aliases
DULLARD
Chromosome Location
Chr.19: 26957150 - 26963304 on Build ARS-UCD1.2
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001046026.1 NP_001039491.1 7 - 8 1025 64
GenBank mRNA BC114677.1 - 7 - 8 1025 64

View Full Product Details