Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
SPON2
Entrez Gene ID
513844
Gene Name
spondin 2
Gene Aliases
-
Chromosome Location
Chr.6: 117253243 - 117256854 on Build ARS-UCD1.2
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001046157.1 NP_001039622.1 4 - 5 814 120
GenBank mRNA BC113291.1 - 4 - 5 814 120

View Full Product Details